siRNA Details
Gene Symbol: LINC00161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026552.1 | 1 | 14 | |||
NR_026553.1 | 1 | 14 | ||||
GenBank mRNA | AY063458.1 | - | 1 | 14 | ||
AY063459.1 | - | 1 | 14 | |||
BC101414.1 | - | 1 | 14 | |||
BC101416.1 | - | 1 | 14 | |||
BC101419.1 | - | 1 | 14 | |||
BC101420.1 | - | 1 | 14 |
