siRNA Details
Gene Symbol: LINC01354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038426.1 | 3 | 2758 | |||
GenBank mRNA | BC040195.2 | - | 3 | 2755 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038426.1 | 3 | 2758 | |||
GenBank mRNA | BC040195.2 | - | 3 | 2755 |