siRNA Details
| Gene Symbol: HOXA10-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046609.1 | 2 | 311 | |||
| GenBank mRNA | BM549446.1 | - | 2 | 282 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ923616.1 | - | 1 | 461 | Chr. 7 - 27169072 - 27171505 [+] on Build GRCh38 | |

HOXA10-AS
Genomic Map