siRNA Details
Gene Symbol: C10orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039380.3 | NP_001034469.2 | 2 | 2055 | ||
XM_017015895.1 | XP_016871384.1 | 3 | 1186 | |||
GenBank mRNA | AK055129.1 | - | 2 | 2033 | ||
AK311351.1 | - | 373 |
