siRNA Details
| Gene Symbol: LOC101927131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110907.1 | 1 | 172 | |||
| GenBank mRNA | BC035158.1 | - | 1 | 175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927131| Gene Symbol: LOC101927131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110907.1 | 1 | 172 | |||
| GenBank mRNA | BC035158.1 | - | 1 | 175 | ||