siRNA Details
| Gene Symbol: C7orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024607.2 | NP_001019778.1 | 2 | 359 | ||
| GenBank mRNA | BG211856.1 | - | 2 | 360 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7orf66| Gene Symbol: C7orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024607.2 | NP_001019778.1 | 2 | 359 | ||
| GenBank mRNA | BG211856.1 | - | 2 | 360 | ||