siRNA Details
| Gene Symbol: SLC16A12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120614.1 | 3 | 462 | |||
| GenBank mRNA | BX089497.1 | - | 3 | 443 | ||
| DB088249.1 | - | 3 | 462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A12-AS1| Gene Symbol: SLC16A12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120614.1 | 3 | 462 | |||
| GenBank mRNA | BX089497.1 | - | 3 | 443 | ||
| DB088249.1 | - | 3 | 462 | |||