siRNA Details
Gene Symbol: LOC101927416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121663.1 | 1 | 178 | |||
NR_121664.1 | 1 | 178 | ||||
GenBank mRNA | BC041457.1 | - | 1 | 136 | ||
DB198681.1 | - | 1 | 178 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121663.1 | 1 | 178 | |||
NR_121664.1 | 1 | 178 | ||||
GenBank mRNA | BC041457.1 | - | 1 | 136 | ||
DB198681.1 | - | 1 | 178 |