siRNA Details
| Gene Symbol: IQCH-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040051.1 | 6 | 1446 | |||
| NR_040052.1 | 6 | 1242 | ||||
| GenBank mRNA | BC047537.1 | - | 6 | 1242 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX451373.1 | - | 2 | 424 | Chr. 15 - 67407629 - 67421391 [-] on Build GRCh38 | |

IQCH-AS1
Genomic Map