siRNA Details
| Gene Symbol: LINC01487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125399.1 | 2 | 99 | |||
| GenBank mRNA | BC037382.1 | - | 1 | 598 | ||
| BG772196.1 | - | 1 | 603 | |||
| BU570602.1 | - | 2 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01487| Gene Symbol: LINC01487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125399.1 | 2 | 99 | |||
| GenBank mRNA | BC037382.1 | - | 1 | 598 | ||
| BG772196.1 | - | 1 | 603 | |||
| BU570602.1 | - | 2 | 99 | |||