siRNA Details
| Gene Symbol: LOC101928233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131192.1 | 2 | 436 | |||
| GenBank mRNA | BC041455.1 | - | 2 | 399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928233| Gene Symbol: LOC101928233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131192.1 | 2 | 436 | |||
| GenBank mRNA | BC041455.1 | - | 2 | 399 | ||