siRNA Details
| Gene Symbol: LINC01505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104145.1 | 5 | 1304 | |||
| GenBank mRNA | AK093363.1 | - | 5 | 1304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01505| Gene Symbol: LINC01505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104145.1 | 5 | 1304 | |||
| GenBank mRNA | AK093363.1 | - | 5 | 1304 | ||