siRNA Details
| Gene Symbol: NAMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102270.1 | 1 | 64 | |||
| NR_102271.1 | 1 | 64 | ||||
| GenBank mRNA | DQ673922.1 | - | 1 | 34 | ||
| DQ673923.1 | - | 1 | 34 | |||
| DQ673924.1 | - | 1 | 34 | |||
| DQ673925.1 | - | 1 | 33 | |||
| DQ673926.1 | - | 1 | 34 | |||
| DQ673963.1 | - | 1 | 600 | |||
| DQ673965.1 | - | 1 | 18 | |||
| DQ673966.1 | - | 1 | 35 | |||

NAMA
Genomic Map