siRNA Details
Gene Symbol: NAMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102270.1 | 10 | 2097 | |||
NR_102271.1 | 6 | 1545 | ||||
GenBank mRNA | DQ673922.1 | - | 10 | 2066 | ||
DQ673929.1 | - | 2 | 201 | |||
DQ673934.1 | - | 3 | 326 |
