siRNA Details
| Gene Symbol: PROSER2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038222.1 | 6 | 3136 | |||
| GenBank mRNA | AK056703.1 | - | 6 | 3125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PROSER2-AS1| Gene Symbol: PROSER2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038222.1 | 6 | 3136 | |||
| GenBank mRNA | AK056703.1 | - | 6 | 3125 | ||