siRNA Details
| Gene Symbol: LINC01574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108030.1 | 2 | 519 | |||
| GenBank mRNA | CK906254.1 | - | 1 | 494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01574| Gene Symbol: LINC01574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108030.1 | 2 | 519 | |||
| GenBank mRNA | CK906254.1 | - | 1 | 494 | ||