siRNA Details
Gene Symbol: LOC101927495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110189.1 | 4 | 1002 | |||
GenBank mRNA | BC040894.1 | - | 4 | 1002 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110189.1 | 4 | 1002 | |||
GenBank mRNA | BC040894.1 | - | 4 | 1002 |