siRNA Details
Gene Symbol: DNAJC27-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034113.1 | 3 | 3098 | |||
GenBank mRNA | AK127070.1 | - | 1 | 1716 | ||
CR933670.1 | - | 3 | 3083 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DNAJC27-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034113.1 | 3 | 3098 | |||
GenBank mRNA | AK127070.1 | - | 1 | 1716 | ||
CR933670.1 | - | 3 | 3083 |