siRNA Details
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521374.2 | XP_011519676.1 | 28 | 5210 | ||
XM_011521378.2 | XP_011519680.1 | 27 | 5051 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521374.2 | XP_011519676.1 | 28 | 5210 | ||
XM_011521378.2 | XP_011519680.1 | 27 | 5051 |