siRNA Details
| Gene Symbol: LINC01204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104644.1 | 3 | 406 | |||
| NR_104645.1 | 3 | 406 | ||||
| GenBank mRNA | AK127697.1 | - | 3 | 382 | ||
| BC040858.1 | - | 3 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01204| Gene Symbol: LINC01204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104644.1 | 3 | 406 | |||
| NR_104645.1 | 3 | 406 | ||||
| GenBank mRNA | AK127697.1 | - | 3 | 382 | ||
| BC040858.1 | - | 3 | 406 | |||