siRNA Details
| Gene Symbol: MIR4300HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120571.1 | 5 | 786 | |||
| GenBank mRNA | BC041900.1 | - | 5 | 786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR4300HG| Gene Symbol: MIR4300HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120571.1 | 5 | 786 | |||
| GenBank mRNA | BC041900.1 | - | 5 | 786 | ||