siRNA Details
| Gene Symbol: LOC643072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110587.1 | 2 | 338 | |||
| NR_110588.1 | 2 | 335 | ||||
| GenBank mRNA | DA209039.1 | - | 2 | 327 | ||
| HY006825.1 | - | 2 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643072| Gene Symbol: LOC643072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110587.1 | 2 | 338 | |||
| NR_110588.1 | 2 | 335 | ||||
| GenBank mRNA | DA209039.1 | - | 2 | 327 | ||
| HY006825.1 | - | 2 | 339 | |||