siRNA Details
Gene Symbol: LINC01142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026957.1 | 4 | 1275 | |||
GenBank mRNA | BC033381.2 | - | 3 | 1205 | ||
BC125144.1 | - | 2 | 672 | |||
BX116913.1 | - | 3 | 300 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026957.1 | 4 | 1275 | |||
GenBank mRNA | BC033381.2 | - | 3 | 1205 | ||
BC125144.1 | - | 2 | 672 | |||
BX116913.1 | - | 3 | 300 |