siRNA Details
| Gene Symbol: LINC01142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026957.1 | 2 | 161 | |||
| GenBank mRNA | BC033381.2 | - | 1 | 91 | ||
| BX116913.1 | - | 2 | 161 | |||
| DB058759.1 | - | 2 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01142| Gene Symbol: LINC01142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026957.1 | 2 | 161 | |||
| GenBank mRNA | BC033381.2 | - | 1 | 91 | ||
| BX116913.1 | - | 2 | 161 | |||
| DB058759.1 | - | 2 | 111 | |||