siRNA Details
| Gene Symbol: LOC100507291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121608.1 | 3 | 289 | |||
| NR_121609.1 | 3 | 289 | ||||
| GenBank mRNA | BX098847.1 | - | 3 | 283 | ||
| DA364121.1 | - | 3 | 252 | |||
| DB034152.1 | - | 3 | 289 | |||
| DB446595.1 | - | 3 | 290 | |||

LOC100507291
Genomic Map