siRNA Details
| Gene Symbol: LOC101928894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120565.1 | 3 | 508 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM478787.1 | - | 3 | 532 | Chr. 11 - 46171947 - 46177066 [+] on Build GRCh38 | |

LOC101928894
Genomic Map