siRNA Details
Gene Symbol: LOC100507654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC034289.1 | - | 4 | 1063 |
Gene Symbol: GGACT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521129.2 | XP_011519431.1 | 1 | 860 |
