siRNA Details
| Gene Symbol: ANO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | HY053628.1 | - | 3 | 429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANO1-AS2| Gene Symbol: ANO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | HY053628.1 | - | 3 | 429 | ||