siRNA Details
| Gene Symbol: UTAT33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110227.1 | 4 | 493 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR739107.1 | - | 4 | 493 | Chr. 2 - 105097060 - 105102944 [-] on Build GRCh38 | |

UTAT33
Genomic Map