siRNA Details
| Gene Symbol: CYB561D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111913.1 | 1 | 95 | |||
| NR_111914.1 | 1 | 95 | ||||
| GenBank mRNA | BM704874.1 | - | 1 | 95 | ||
| CT000022.1 | - | 1 | 44 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM925473.1 | - | 73 | Chr. - - [-] on Build | ||

CYB561D2
Genomic Map