siRNA Details
| Gene Symbol: LOC101928002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110072.1 | 4 | 1318 | |||
| GenBank mRNA | BC042465.1 | - | 4 | 1318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928002| Gene Symbol: LOC101928002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110072.1 | 4 | 1318 | |||
| GenBank mRNA | BC042465.1 | - | 4 | 1318 | ||