siRNA Details
| Gene Symbol: LINC00934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024246.2 | 2 | 415 | |||
| GenBank mRNA | AK057497.1 | - | 2 | 415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00934| Gene Symbol: LINC00934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024246.2 | 2 | 415 | |||
| GenBank mRNA | AK057497.1 | - | 2 | 415 | ||