siRNA Details
| Gene Symbol: CA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121633.1 | 2, 3 | 197 | |||
| GenBank mRNA | AW611902.1 | - | 1, 2 | 63 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CA3-AS1| Gene Symbol: CA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121633.1 | 2, 3 | 197 | |||
| GenBank mRNA | AW611902.1 | - | 1, 2 | 63 | ||