siRNA Details
Gene Symbol: CA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121630.1 | 2 | 346 | |||
NR_121631.1 | 2 | 118 | ||||
NR_121632.1 | 2 | 135 | ||||
NR_121633.1 | 2 | 135 | ||||
GenBank mRNA | BM713112.1 | - | 1 | 26 | ||
BM717037.1 | - | 1 | 26 | |||
DA940436.1 | - | 2 | 118 | |||
DB177224.1 | - | 2 | 205 | |||
DB196812.1 | - | 2 | 346 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB177284.1 | - | 2 | 205 | Chr. 8 - 85441947 - 85464774 [-] on Build GRCh38 |
