siRNA Details
Gene Symbol: TMEM220-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073455.1 | 5 | 619 | |||
NR_073456.1 | 4 | 523 | ||||
GenBank mRNA | BQ694074.1 | - | 4 | 489 | ||
BQ694934.1 | - | 4 | 477 | |||
DA747858.1 | - | 3 | 489 |
