siRNA Details
| Gene Symbol: LOC101927815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125423.1 | 5 | 912 | |||
| GenBank mRNA | BC030605.2 | - | 5 | 913 | ||
| BC045738.1 | - | 5 | 913 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927815| Gene Symbol: LOC101927815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125423.1 | 5 | 912 | |||
| GenBank mRNA | BC030605.2 | - | 5 | 913 | ||
| BC045738.1 | - | 5 | 913 | |||