siRNA Details
| Gene Symbol: MIR670HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131246.1 | 1 | 94 | |||
| GenBank mRNA | BC031305.2 | - | 1 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR670HG| Gene Symbol: MIR670HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131246.1 | 1 | 94 | |||
| GenBank mRNA | BC031305.2 | - | 1 | 94 | ||