siRNA Details
Gene Symbol: LHFPL3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034141.1 | 3 | 341 | |||
NR_034142.1 | 3 | 341 | ||||
GenBank mRNA | BU161581.1 | - | 3 | 341 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LHFPL3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034141.1 | 3 | 341 | |||
NR_034142.1 | 3 | 341 | ||||
GenBank mRNA | BU161581.1 | - | 3 | 341 |