siRNA Details
| Gene Symbol: LINC01547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027129.1 | 2 | 378 | |||
| GenBank mRNA | AF380178.1 | - | 2 | 310 | ||
| AK127059.1 | - | 1 | 2288 | |||
| AY040088.1 | - | 2 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01547| Gene Symbol: LINC01547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027129.1 | 2 | 378 | |||
| GenBank mRNA | AF380178.1 | - | 2 | 310 | ||
| AK127059.1 | - | 1 | 2288 | |||
| AY040088.1 | - | 2 | 310 | |||