siRNA Details
| Gene Symbol: LINC01547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027128.1 | 4 | 1401 | |||
| GenBank mRNA | BC101235.2 | - | 1 | 639 | ||
| BC101236.2 | - | 1 | 639 | |||
| BC101237.1 | - | 1 | 639 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01547| Gene Symbol: LINC01547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027128.1 | 4 | 1401 | |||
| GenBank mRNA | BC101235.2 | - | 1 | 639 | ||
| BC101236.2 | - | 1 | 639 | |||
| BC101237.1 | - | 1 | 639 | |||