siRNA Details
| Gene Symbol: CACNA1C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045725.1 | 4 | 2201 | |||
| GenBank mRNA | AK093746.1 | - | 4 | 2161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CACNA1C-AS1| Gene Symbol: CACNA1C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045725.1 | 4 | 2201 | |||
| GenBank mRNA | AK093746.1 | - | 4 | 2161 | ||