siRNA Details
| Gene Symbol: SEC24B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039978.1 | 3 | 193 | |||
| GenBank mRNA | BC009800.1 | - | 3 | 193 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC24B-AS1| Gene Symbol: SEC24B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039978.1 | 3 | 193 | |||
| GenBank mRNA | BC009800.1 | - | 3 | 193 | ||