siRNA Details
| Gene Symbol: LOC100996286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121627.1 | 3 | 796 | |||
| NR_121628.1 | 3 | 460 | ||||
| GenBank mRNA | BC040914.1 | - | 3 | 796 | ||
| CB986668.1 | - | 463 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996286| Gene Symbol: LOC100996286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121627.1 | 3 | 796 | |||
| NR_121628.1 | 3 | 460 | ||||
| GenBank mRNA | BC040914.1 | - | 3 | 796 | ||
| CB986668.1 | - | 463 | ||||