siRNA Details
| Gene Symbol: LOC51145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122032.1 | 2 | 589 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX095101.1 | - | 2 | 587 | Chr. 9 - 125743770 - 125745927 [-] on Build GRCh38 | |

LOC51145
Genomic Map