siRNA Details
| Gene Symbol: C1orf140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024236.1 | 2 | 1691 | |||
| GenBank mRNA | AK128488.1 | - | 2 | 1691 | ||
| BC140931.1 | - | 2 | 1524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf140| Gene Symbol: C1orf140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024236.1 | 2 | 1691 | |||
| GenBank mRNA | AK128488.1 | - | 2 | 1691 | ||
| BC140931.1 | - | 2 | 1524 | |||