siRNA Details
| Gene Symbol: DEAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104273.1 | 2 | 160 | |||
| GenBank mRNA | EF212178.1 | - | 2 | 160 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEAR| Gene Symbol: DEAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104273.1 | 2 | 160 | |||
| GenBank mRNA | EF212178.1 | - | 2 | 160 | ||