siRNA Details
| Gene Symbol: LINC01107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037809.1 | 3 | 1193 | |||
| GenBank mRNA | AK057376.1 | - | 3 | 748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01107| Gene Symbol: LINC01107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037809.1 | 3 | 1193 | |||
| GenBank mRNA | AK057376.1 | - | 3 | 748 | ||