siRNA Details
| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002770.1 | 1 | 3420 | |||
| GenBank mRNA | AF469201.1 | - | 2 | 2439 | ||
| AF469202.1 | - | 1 | 3420 | |||
| AF469203.1 | - | 2 | 951 | |||
| AF469206.1 | - | 3 | 918 | |||
| AF469207.1 | - | 3 | 825 | |||
| AF469208.1 | - | 2 | 650 | |||

DIO3OS
Genomic Map