siRNA Details
Gene Symbol: DIO3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF305836.1 | - | 4 | 997 | ||
AF469199.1 | - | 3 | 820 | |||
AF469200.1 | - | 4 | 666 | |||
AF469205.1 | - | 1 | 51 | |||
BC037351.1 | - | 1 | 3061 | |||
BC065701.1 | - | 3 | 1835 | |||
BC111049.1 | - | 2 | 1652 |
