siRNA Details
| Gene Symbol: LOC101926955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110003.1 | 4 | 3841 | |||
| GenBank mRNA | BC052952.1 | - | 4 | 3841 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926955| Gene Symbol: LOC101926955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110003.1 | 4 | 3841 | |||
| GenBank mRNA | BC052952.1 | - | 4 | 3841 | ||